Få mere at vide
p-Toluic Acid-d7, TRC
Artikelnummer. 30452074
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30452074 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452074 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53612850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Toluic Acid-d7
CAS 1219798-76-1
Molekylær formel C8 D7 H O2
Mængde 50 mg
Synonym Benzoic-2,3,5,6-d4 acid','4-(methyl-d3)- (ACI), 4-(Methyl-d3)benzoic-2,3,5,6-d4 acid (ACI), p-Toluic-α,α,α,2,3,5,6-d7 acid, 4-Methylbenzoic acid D7, p-Toluic acid-d7
InChI formel InChI=1S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C(=O)O)C([2H])([2H])[2H]
IUPAC navn 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 143.191
Formel vægt 143.096