Få mere at vide
p-Toluic-2,3,5,6-d4 Acid, CDN
Artikelnummer. 30317651
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317651 0.25 g
30312935 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317651 Leverandør CDN Leverandørnr. CDND59540.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylbenzoic-2,3,5,6-D4 Acid
CAS 1219798-71-6
Molekylær formel C8 D4 H4 O2
Mængde 0.25 g
Synonym Benzoic-2,3,5,6-d4 acid, 4-methyl- (ACI), 4-Methylbenzoic-2,3,5,6-d4 acid (ACI), p-Toluic-2,3,5,6-d4 acid, 4-Methylbenzoic-2,3,5,6-D4 acid, p-Toluic acid-d4
InChI formel InChI=1 S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5 H,1H3,(H,9,10)/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1C
IUPAC navn 2,3,5,6-tetradeuterio-4-methylbenzoic acid
Molekylvægt (g/mol) 140.173
Formel vægt 140.078 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity