Få mere at vide
p-Tolualdehyde-d7 (2,3,5,6-d4; methyl-d3), CDN
Artikelnummer. 30319209
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319209 0.25 g
30315803 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319209 Leverandør CDN Leverandørnr. CDND64970.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylbenzaldehyde-D7
CAS 1219805-23-8
Molekylær formel C8 D7 H O
Mængde 0.25 g
Synonym p-Tolualdehyde-d7, 4-Methylbenzaldehyde-D7, 4-(Methyl-d3)benzaldehyde-2,3,5,6-d4, 4-Tolualdehyde-d7
InChI formel InChI=1 S/C8H8O/c1-7-2-4-8(6-9)5-3-7/h2-6 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C=O)C([2 H])([2 H])[2 H]
IUPAC navn 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzaldehyde
Molekylvægt (g/mol) 127.192
Formel vægt 127.101 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity