Få mere at vide
p-SCN-Bn-deferoxamine, TRC
Artikelnummer. 30008542
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008542 50 mg
30003916 25 mg
30005048 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008542 Leverandør Toronto Research Chemicals Leverandørnr. TRCS20012550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1222468-90-7
Molekylær formel C33H52N8O8S2
Molekylvægt (g/mol) 752.94
Synonym N4-[5-[[4-[[5-(Acetylhydroxyamino)pentyl]amino]-1,4-dioxobutyl]hydroxyamino]pentyl]-N1-hydroxy-N1-[5-[[[(4-isothiocyanatophenyl)amino]thioxomethyl]amino]pentyl]butanediamide, P-Isothiocyanatobenzyl-desferrioxamine
IUPAC navn N-[5-[acetyl(hydroxy)amino]pentyl]-N'-hydroxy-N'-[5-[[4-[hydroxy-[5-[(4-isothiocyanatophenyl)carbamothioylamino]pentyl]amino]-4-oxobutanoyl]amino]pentyl]butanediamide
SMIL CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=S)Nc1ccc(cc1)N=C=S
Tekniske data
Kemisk navn eller materiale p-SCN-Bn-deferoxamine
Mængde 50 mg
InChI formel InChI=1S/C33H52N8O8S2/c1-26(42)39(47)22-8-2-5-19-34-29(43)15-17-31(45)40(48)23-9-3-6-20-35-30(44)16-18-32(46)41(49)24-10-4-7-21-36-33(51)38-28-13-11-27(12-14-28)37-25-50/h11-14,47-49H,2-10,15-24H2,1H3,(H,34,43)(H,35,44)(H2,36,38,51)
Formel vægt 752.335