Få mere at vide
p-Phthalamidine Dihydrochloride, TRC
Artikelnummer. 17922475
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17922475 5 g
17973705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17922475 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3824705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14401-56-0
Molekylær formel C8 H10 N4 . 2 Cl H
Molekylvægt (g/mol) 235.114
Synonym 1,4-Benzenedicarboximidamide Dihydrochloride, Terephthalamidine Dihydrochloride
IUPAC navn benzene-1,4-dicarboxamidine;dihydrochloride
SMIL Cl.Cl.NC(=N)c1ccc(cc1)C(=N)N
Tekniske data
Kemisk navn eller materiale p-Phthalamidine Dihydrochloride
Mængde 5 g
InChI formel InChI=1S/C8H10N4.2ClH/c9-7(10)5-1-2-6(4-3-5)8(11)12;;/h1-4H,(H3,9,10)(H3,11,12);2*1H
Formel vægt 234.0439