Få mere at vide
p-Phenetidine-d5 Hydrochloride, TRC
Artikelnummer. 30421372
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421372 1 mg
30416796 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421372 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2963021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794754-45-2
Molekylær formel C8H7D5ClNO
Molekylvægt (g/mol) 178.67
Synonym 4-(Ethoxy-d5)benzenamine Hydrochloride, 1-Amino-4-(ethoxy-d5)benzene Hydrochloride, 4-Aminophenetole-d5 Hydrochloride, 4-Ethoxyaniline-d5 Hydrochloride, 4-(Ethoxy-d5)benzenamine Hydrochloride, 4-(Ethoxy-d5)phenylamine Hydrochloride, NSC 3116-d5 Hydrochloride, p-Aminophenetole-d5 Hydrochloride, p-Ethoxyaniline-d5 Hydrochloride, p-(Ethoxy-d5)phenylamine Hydrochloride, p-Phenetidin-d5 Hydrochloride
IUPAC navn 4-(1,1,2,2,2-pentadeuterioethoxy)aniline;hydrochloride
SMIL NC1=CC=C(OC([2H])([2H])C([2H])([2H])[2H])C=C1.Cl
Tekniske data
Kemisk navn eller materiale p-Phenetidine-d5 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C8H11NO.ClH/c1-2-10-8-5-3-7(9)4-6-8;/h3-6H,2,9H2,1H3;1H/i1D3,2D2;
Formel vægt 178.0921