Få mere at vide
p-Nitrophenylthiol Acetate, TRC
Artikelnummer. 30467873
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467873 500 mg
30464894 1 g
30465144 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30467873 Leverandør Toronto Research Chemicals Leverandørnr. TRCN505900500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15119-62-7
Molekylær formel C8 H7 N O3 S
Molekylvægt (g/mol) 197.21
Synonym S-(4-Nitrophenyl) Ester Ethanethioic Acid, S-(p-Nitrophenyl) Ester Thioacetic Acid, p-Nitrobenzenethiol Acetate, 4-Nitrophenyl Thiolacetate, S-(4-Nitrophenyl)thioacetate, S-(p-Nitrophenyl) Thioacetate, S-4-Nitrophenyl Ethanethioate
IUPAC navn S-(4-nitrophenyl) ethanethioate
SMIL CC(=O)Sc1ccc(cc1)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale p-Nitrophenylthiol Acetate
Mængde 500 mg
InChI formel InChI=1S/C8H7NO3S/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3
Formel vægt 197.0147