Få mere at vide
p-Nitrophenyl Beta-D-Lactopyranoside, TRC
Artikelnummer. 30479094
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479094 500 mg
30474567 100 mg
30469539 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479094 Leverandør Toronto Research Chemicals Leverandørnr. TRCN503850500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4419-94-7
Molekylær formel C18 H25 N O13
Molekylvægt (g/mol) 463.39
Synonym β-D-Glucopyranoside, 4-nitrophenyl 4-O-β-D-galactopyranosyl-, Lactoside, p-nitrophenyl, β-D- (6CI,8CI), 4-Nitrophenyl 4-O-β-D-galactopyranosyl-β-D-glucopyranoside, 4-Nitrophenyl β-lactoside, p-Nitrophenyl β-D-lactoside, p-Nitrophenyl-β-lactoside
IUPAC navn (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](Oc3ccc(cc3)[N+](=O)[O-])O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale p-Nitrophenyl beta-D-Lactopyranoside
Mængde 500 mg
InChI formel InChI=1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11+,12+,13-,14-,15-,16-,17-,18+/m1/s1
Formel vægt 463.1326