missing translation for 'onlineSavingsMsg'
Få mere at vide

p-Nitrophenyl Beta-D-Lactopyranoside Heptaacetate, TRC

Artikelnummer. 30483547
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483547 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30483547 Leverandør Toronto Research Chemicals Leverandørnr. TRCN50386025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale p-Nitrophenyl beta-D-Lactopyranoside Heptaacetate
CAS 84034-75-3
Molekylær formel C32H39NO20
Mængde 25 mg
Synonym 4-Nitrophenyl 4-O-(2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranosyl)-beta-D-glucopyranoside 2,3,6-Triacetate
InChI formel InChI=1S/C32H39NO20/c1-14(34)43-12-23-25(45-16(3)36)27(46-17(4)37)30(49-20(7)40)32(52-23)53-26-24(13-44-15(2)35)51-31(29(48-19(6)39)28(26)47-18(5)38)50-22-10-8-21(9-11-22)33(41)42/h8-11,23-32H,12-13H2,1-7H3/t23-,24-,25+,26-,27+,28+,29-,30-,31-,32+/m1/s1
SMIL CC(=O)OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1O[C@@H]3O[C@H](COC(=O)C)[C@H](OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C
IUPAC navn [(2R,3R,4S,5R,6S)-4,5-diacetyloxy-6-(4-nitrophenoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
Molekylvægt (g/mol) 757.65
Formel vægt 757.2065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.