missing translation for 'onlineSavingsMsg'
Få mere at vide

p-Nitrophenyl Beta-D-Cellobioside, TRC

Artikelnummer. 30475408
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475408 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30475408 Leverandør Toronto Research Chemicals Leverandørnr. TRCN50309010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale p-Nitrophenyl beta-D-Cellobioside
CAS 3482-57-3
Molekylær formel C18 H25 N O13
Mængde 10 mg
Synonym β-D-Glucopyranoside','4-nitrophenyl 4-O-β-D-glucopyranosyl-, Cellobioside','p-nitrophenyl (7CI), Cellobioside','p-nitrophenyl','β-D- (8CI), 4-Nitrophenyl 4-O-β-D-glucopyranosyl-β-D-glucopyranoside, p-Nitrophenyl cellobioside, p-Nitrophenyl β-D-cellobioside, p-Nitrophenyl β-cellobioside
InChI formel InChI=1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12+,13-,14-,15-,16-,17-,18+/m1/s1
SMIL OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](Oc3ccc(cc3)[N+](=O)[O-])O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC navn (2S,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 463.39
Formel vægt 463.1326
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.