Få mere at vide
p-Nitrophenyl alpha-D-Galactopyranoside, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Nitrophenyl-alpha-D-galactopyranoside |
| CAS | 7493-95-0 |
| Molekylær formel | C12 H15 N O8 |
| Mængde | 1 g |
| Synonym | α-D-Galactopyranoside','4-nitrophenyl, Galactopyranoside','p-nitrophenyl','α-D- (8CI), 4-Nitrophenyl α-D-galactopyranoside, 4-Nitrophenyl α-D-galactoside, NSC 89286, PNPG, p-Nitrophenyl α-D-galactopyranoside, p-Nitrophenyl α-D-galactoside, p-Nitrophenyl α-galactopyranoside, p-Nitrophenyl α-galactoside |
| InChI formel | InChI=1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9+,10+,11-,12+/m1/s1 |
| SMIL | OC[C@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@@H](O)[C@H]1O |
| IUPAC navn | (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| Molekylvægt (g/mol) | 301.25 |
| Formel vægt | 301.0798 |