Få mere at vide
p-Nitrophenyl 4,6-Benzylidene-Alpha-D-glucopyranoside, TRC
Artikelnummer. 30473888
Skift visning
Mængde:
500 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30473888 500 mg -
30467706 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473888 Leverandør Toronto Research Chemicals Leverandørnr. TRCN503055500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Nitrophenyl 4,6-Benzylidene-a-D-glucopyranoside
CAS 85906-27-0
Molekylær formel C19 H19 N O8
Mængde 500 mg
InChI formel InChI=1S/C19H19NO8/c21-15-16(22)19(26-13-8-6-12(7-9-13)20(23)24)27-14-10-25-18(28-17(14)15)11-4-2-1-3-5-11/h1-9,14-19,21-22H,10H2/t14-,15-,16-,17-,18?,19+/m1/s1
SMIL O[C@H]1[C@@H](Oc2ccc(cc2)[N+](=O)[O-])O[C@@H]3COC(O[C@H]3[C@@H]1O)c4ccccc4
IUPAC navn (4aR,6R,7R,8R,8aS)-6-(4-nitrophenoxy)-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
Molekylvægt (g/mol) 389.36
Formel vægt 389.1111