Få mere at vide
p-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(Beta-D-galactopyranosyl)-Beta-D-glucopyranoside, TRC
Artikelnummer. 30483661
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30483661 5 mg -
30477274 1 mg -
30466601 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483661 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5012205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57467-13-7
Molekylær formel C20 H28 N2 O13
Molekylvægt (g/mol) 504.44
IUPAC navn N-[(2S,3R,4R,5S,6R)-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
SMIL CC(=O)N[C@H]1[C@H](Oc2ccc(cc2)[N+](=O)[O-])O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O
Tekniske data
Kemisk navn eller materiale p-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(beta-D-galactopyranosyl)-beta-D-glucopyranoside
Mængde 5 mg
InChI formel InChI=1S/C20H28N2O13/c1-8(25)21-13-18(35-20-17(29)16(28)14(26)11(6-23)34-20)15(27)12(7-24)33-19(13)32-10-4-2-9(3-5-10)22(30)31/h2-5,11-20,23-24,26-29H,6-7H2,1H3,(H,21,25)/t11-,12-,13-,14+,15-,16+,17-,18-,19-,20+/m1/s1
Formel vægt 504.1591