missing translation for 'onlineSavingsMsg'
Få mere at vide

p-Nitrobenzoyl-2,3,4-tri-O-benzyl-Alpha,Beta-L-fucopyranose, TRC

Artikelnummer. 30474291
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474291 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30474291 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCN495000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale p-Nitrobenzoyl-2,3,4-tri-O-benzyl-a,b-L-fucopyranose
CAS 151909-88-5
Molekylær formel C34 H33 N O8
Mængde 250 mg
InChI formel InChI=1S/C34H33NO8/c1-24-30(39-21-25-11-5-2-6-12-25)31(40-22-26-13-7-3-8-14-26)32(41-23-27-15-9-4-10-16-27)34(42-24)43-33(36)28-17-19-29(20-18-28)35(37)38/h2-20,24,30-32,34H,21-23H2,1H3/t24-,30+,31+,32-,34?/m0/s1
SMIL C[C@@H]1OC(OC(=O)c2ccc(cc2)[N+](=O)[O-])[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@@H]1OCc5ccccc5
IUPAC navn [(3S,4R,5R,6S)-6-methyl-3,4,5-tris(phenylmethoxy)oxan-2-yl] 4-nitrobenzoate
Molekylvægt (g/mol) 583.63
Formel vægt 583.2206
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.