missing translation for 'onlineSavingsMsg'
Få mere at vide

p-Methyl-cinnamoyl Azide, TRC

Artikelnummer. 30477804
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30477804 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2945152G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale p-Methyl-cinnamoyl Azide
CAS 24186-38-7
Molekylær formel C10H9N3O
Mængde 2 g
Synonym 3-(4-Methylphenyl)-2-propenoyl Azide
InChI formel InChI=1S/C10H9N3O/c1-8-2-4-9(5-3-8)6-7-10(14)12-13-11/h2-7H,1H3/b7-6+
SMIL Cc1ccc(\C=C\C(=O)N=[N+]=[N-])cc1
IUPAC navn (E)-3-(4-methylphenyl)prop-2-enoyl azide
Molekylvægt (g/mol) 187.2
Formel vægt 187.0746
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.