Få mere at vide
p-Ethylphenol-d9, TRC
Artikelnummer. 30371024
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371024 50 mg
30382692 100 mg
30371677 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371024 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92573250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2241872-74-0
Molekylær formel C8HD9O
Molekylvægt (g/mol) 131.22
Synonym Phen-2,3,5,6-d4-ol, 4-(ethyl-1,1,2,2,2-d5)- (ACI), 4-(Ethyl-1,1,2,2,2-d5)phen-2,3,5,6-d4-ol (ACI), 4-Ethylphenol-d9, 4-Ethylphenol-D9, p-Ethylphenol-d9
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1O)C([2H])([2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale p-Ethylphenol-d9
Mængde 50 mg
InChI formel InChI=1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3/i1D3,2D2,3D,4D,5D,6D
Formel vægt 131.1297