Få mere at vide
p-Cyanoacetophenone-d4, TRC
Artikelnummer. 30358629
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30358629 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358629 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98750710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Cyanoacetophenone-d4
CAS 1794964-28-5
Molekylær formel C9H3D4NO
Mængde 10 mg
Synonym 1-Acetyl-4-benzonitrile-d4, 4-Acetylbenzonitrile-d4, 4-Cyanoacetophenone-d4, 4-Cyanophenyl-d4 Methyl Ketone, 4'-Cyanoacetophenone-d4, Methyl p-Cyanophenyl-d4 Ketone, p-Acetylbenzonitrile-d4, p-Cyanoacetophenone-d4, p-Cyanophenyl-d4 Methyl Ketone
InChI formel InChI=1S/C9H7NO/c1-7(11)9-4-2-8(6-10)3-5-9/h2-5H,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(C(=O)C)c([2H])c([2H])c1C#N
IUPAC navn 4-acetyl-2,3,5,6-tetradeuteriobenzonitrile
Molekylvægt (g/mol) 149.18
Formel vægt 149.0779