Få mere at vide
p-Cresol-2,3,5,6-d4,OD, CDN
Artikelnummer. 30319322
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319322 0.5 g 0.5g
30317309 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319322 Leverandør CDN Leverandørnr. CDND18370.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylphenol-2,3,5,6,O-D5
CAS 3646-98-8
Molekylær formel C7 D5 H3 O
Mængde 0.5 g
Synonym p-Cres-2,3,5,6-d4-ol-d (7 CI, 8 CI), 2,3,5,6,O-Pentadeuterio-4-methylphenol, 4-Methylphen-2,3,5,6-d4-ol-d, p-Cresol-d5, p-Cresol-2,3,5,6-d4,OD, 4-Methylphenol-2,3,5,6,O-D5
InChI formel InChI=1 S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8 H,1H3/i2D,3 D,4 D,5 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c(C)c([2 H])c1[2 H]
IUPAC navn 1,2,4,5-tetradeuterio-3-deuteriooxy-6-methylbenzene
Molekylvægt (g/mol) 113.169
Formel vægt 113.089 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity