missing translation for 'onlineSavingsMsg'
Få mere at vide

p-Chlorophenylcyanoguanidine, TRC

Artikelnummer. 30358514
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
25 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30358514 2.5 g 2.5g
30350123 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30358514 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3777352.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1482-62-8
Molekylær formel C8 H7 Cl N4
Molekylvægt (g/mol) 194.62
Synonym Guanidine, N-(4-chlorophenyl)-N'-cyano-, Guanidine, 1-(p-chlorophenyl)-3-cyano- (6CI,7CI,8CI), N-(4-Chlorophenyl)-N'-cyanoguanidine, 1-(p-Chlorophenyl)-3-cyanoguanidine, 1-Cyano-3-(4-chlorophenyl)guanidine, N-(p-Chlorophenyl)dicyandiamide, N1-(4-Chlorophenyl)-N3-cyanoguanidine, NSC 74463, p-Chlorophenylcyanoguanidine, p-Chlorophenyldicyandiamide, Proguanil Hydrochloride Imp. E (EP)
IUPAC navn 1-(4-chlorophenyl)-3-cyanoguanidine
SMIL Clc1ccc(NC(=N)NC#N)cc1

Tekniske data

Kemisk navn eller materiale 1-(4-chlorophenyl)-3-cyanoguanidine
Mængde 2.5 g
InChI formel InChI=1S/C8H7ClN4/c9-6-1-3-7(4-2-6)13-8(11)12-5-10/h1-4H,(H3,11,12,13)
Formel vægt 194.0359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.