missing translation for 'onlineSavingsMsg'
Learn More

P-Cadherin, APC, clone: CSTEM29, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen 17-2237-41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15855353

  • 1012.00 DKK / Pack of 25


Estimated Shipment date
from Supplier 04-02-2019
Add to basket

Description

Description

This monoclonal antibody CSTEM29 reacts with human P-Cadherin, also known as Cadherin-3.Applications Reported: This CSTEM29 antibody has been reported for use in flow cytometric analysis.Applications Tested: This CSTEM29 antibody has been pre-diluted and tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at 5μL (0.25μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647nm; Emission: 660nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

P-Cadherin
Monoclonal
5μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P22223
Cadherin-3, CDH3, PCAD
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CSTEM29
APC
CDH3
Liquid
CDH3
IgG2a, kappa
25 Tests
4°C, store in dark, DO NOT FREEZE!
Primary
1001
Videos