Få mere at vide
p-Bromofluorobenzene-d4, TRC
Artikelnummer. 30333442
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30333442 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333442 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68454750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-Bromofluorobenzene-d4
CAS 50592-31-9
Molekylær formel C6 D4 Br F
Mængde 50 mg
Synonym 3-Bromo-6-fluoro-benzene-1,2,4,5-d4, 4-Bromofluorobenzene-d4
InChI formel InChI=1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(Br)c([2H])c([2H])c1F
IUPAC navn 1-bromo-2,3,5,6-tetradeuterio-4-fluorobenzene
Molekylvægt (g/mol) 179.02
Formel vægt 177.9731