Få mere at vide
p-Azoxyanisole-d6 (O,O-dimethyl-d6), CDN
Artikelnummer. 30323325
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323325 0.25 g
30321477 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323325 Leverandør CDN Leverandørnr. CDND17010.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4'-Dimethoxyazoxybenzene-D6 (dimethoxy-D6)
CAS 34172-21-9
Molekylær formel C14 D6 H8 N2 O3
Mængde 0.25 g
Synonym Diazene, bis[4-(methoxy-d3)phenyl]-, 1-oxide, (1 Z)- (9 CI, ACI), Azoxybenzene, 4,4'-di(methoxy-d3)- (8 CI), Diazene, bis[4-(methoxy-d3)phenyl]-, 1-oxide, (Z)- (ZCI), Methyl-deuterated p-azoxyanisole, p-Azoxyanisole-α,α,α,α',α',α'-d6, 4,4'-Dimethoxyazoxybenzene-D6 (dimethoxy-D6), 4,4'-Dimethoxyazoxybenzene-D6 (dimethoxy-D6), 4,4'-Dimethoxyazoxybenzene-D6 (dimethoxy-D6), 4,4'-Dimethoxyazoxybenzene-D6 (dimethoxy-D6)
InChI formel InChI=1 S/C14H14N2O3/c1-18-13-7-3-11(4-8-13)15-16(17)12-5-9-14(19-2)10-6-12/h3-10 H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])Oc1ccc(cc1)N=[N+]([O-])c2ccc(OC([2 H])([2 H])[2 H])cc2
IUPAC navn oxido-[4-(trideuteriomethoxy)phenyl]-[4-(trideuteriomethoxy)phenyl]iminoazanium
Molekylvægt (g/mol) 264.31
Formel vægt 264.138 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity