Få mere at vide
Tocris Bioscience™ P-3FAX-Neu5Ac
Sialic acid analog; sialyltransferase inhibitor
Brand: Tocris Bioscience™ 5760/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
P-3FAX-Neu5Ac is a cell-permeable sialic acid analog. Deacetylation occurs intracellularly giving rise to a sialyltransferase inhibitor. Abolishes expression of SLex expression on HL-60 cells and reduces E-selectin and P-selectin binding. Prevents metastasis formation in a mouse lung metastasis model.Tekniske data
| 5-(acetylamino)-3,5-dideoxy-3-fluoro-D-erythro-α-L-manno-2-nonulopyranosonic acid methyl ester 2,4,7,8,9-pentaacetate | |
| 117405-58-0 | |
| 10 mg |