Få mere at vide
Ozagrel, MedChemExpress
Artikelnummer. 16498296
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498296 100 mg
16494357 50 mg
16428488 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16498296 Leverandør Medchem Express Leverandørnr. HYB0428
MedChemExpress Ozagrel (OKY-046)is an anti-asthmatic agent and a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM.
Ozagrel (OKY-046)is an anti-asthmatic agent and a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM.
Tekniske data
Kemisk navn eller materiale Ozagrel
CAS 82571-53-7
Farve White
Renhedsgrad noter Research
Molekylær formel C13H12N2O2
Mængde 100 mg
Synonym OKY-046
Opløselighedsinformation DMSO : 50 mg/mL (219.06 mM; Need ultrasonic)
SMIL O=C(O)/C=C/C1=CC=C(CN2C=CN=C2)C=C1
Molekylvægt (g/mol) 228.25
Formel vægt 228.25
Procent renhed 99.96%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation