missing translation for 'onlineSavingsMsg'
Få mere at vide

Oxyresveratrol, TRC

Artikelnummer. 30417653
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417653 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417653 Leverandør Toronto Research Chemicals Leverandørnr. TRCO861920100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale trans-Oxyresveratrol
CAS 29700-22-9
Molekylær formel C14 H12 O4
Mængde 100 mg
Synonym 1,3-Benzenediol','4-[(1E)-2-(3,5-dihydroxyphenyl)ethenyl]-, 1,3-Benzenediol','4-[2-(3,5-dihydroxyphenyl)ethenyl]-','(E)-, 2,3',4,5'-Stilbenetetrol','(E)- (8CI), 4-[(1E)-2-(3,5-Dihydroxyphenyl)ethenyl]-1,3-benzenediol, Oxyresveratrol, trans-Oxyresveratrol, trans-Stilbene-2,4,3',5'-tetrol
InChI formel InChI=1S/C14H12O4/c15-11-4-3-10(14(18)8-11)2-1-9-5-12(16)7-13(17)6-9/h1-8,15-18H/b2-1+
SMIL Oc1ccc(\C=C\c2cc(O)cc(O)c2)c(O)c1
IUPAC navn 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Molekylvægt (g/mol) 244.24
Formel vægt 244.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.