missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Oxyphenonium Bromide, Mikromol™
Beskrivelse
Oxyphenonium Bromide
Specifications
Specifications
| Kemisk navn eller materiale | Oxyphenonium Bromide |
| CAS | 50-10-2 |
| Analyt- eller komponentnavne | Oxyphenonium Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H34 N O3 . Br |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ethanaminium, 2-[(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (1:1), Ethanaminium, 2-[(cyclohexylhydroxyphenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (9CI), Antrenil, Antrenyl, Antrenyl Duplex, Ba-5473, C 5473, Oxifenon, Oxyfenon, Oxyphenon, Oxyphenonium bromide, Spasmodin, Spasmophen, dl-Oxyphenonium bromide, Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, alpha-phenylcyclohexaneglycolate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide alpha-phenylcyclohexaneglycolate (6CI), Cyclohexaneglycolic acid, alpha-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), (+/-)-Oxyphenonium bromide |
| InChI formel | InChI=1S/C21H34NO3.BrH/c1-4-22(3,5-2)16-17-25-20(23)21(24,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6,8-9,12-13,19,24H,4-5,7,10-11,14-17H2,1-3H3;1H/q+1;/p-1 |
| SMIL | [Br-].CC[N+](C)(CC)CCOC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC navn | 2-(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxyethyl-diethyl-methylazanium;bromide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?