Få mere at vide
Oxyphenonium Betaromide, TRC
Artikelnummer. 30417799
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417799 10 mg
30414732 50 mg
30411727 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30417799 Leverandør Toronto Research Chemicals Leverandørnr. TRCO87696810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50-10-2
Molekylær formel C21 H34 N O3 . Br
Molekylvægt (g/mol) 428.4
Synonym Ethanaminium, 2-[(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (1:1), Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, α-phenylcyclohexaneglycolate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide α-phenylcyclohexaneglycolate (6CI), Ethanaminium, 2-[(cyclohexylhydroxyphenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (9CI), Cyclohexaneglycolic acid, α-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), (±)-Oxyphenonium bromide, Antrenil, Antrenyl, Antrenyl Duplex, Ba-5473, C 5473, Oxifenon, Oxyfenon, Oxyphenon, Oxyphenonium bromide, Spasmodin, Spasmophen, dl-Oxyphenonium bromide
IUPAC navn 2-(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxyethyl-diethyl-methylazanium;bromide
SMIL [Br-].CC[N+](C)(CC)CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Oxyphenonium Bromide
Mængde 10 mg
InChI formel InChI=1S/C21H34NO3.BrH/c1-4-22(3,5-2)16-17-25-20(23)21(24,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6,8-9,12-13,19,24H,4-5,7,10-11,14-17H2,1-3H3;1H/q+1;/p-1
Formel vægt 427.1722