Få mere at vide
Oxyphenbutazone Monohydrate, LoGiCal
Artikelnummer. 30461663
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461663 10 mg
1 options
Artikelnummer. 30461663 Leverandør LoGiCal Leverandørnr. LGCFOR0933.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxyphenbutazone Monohydrate
CAS 7081-38-1
Analyt- eller komponentnavne Oxyphenbutazone Monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C19 H20 N2 O3 . H2 O
Certificeringer/compliance ISO 9001
Synonym 4-Butyl-1-(4-hydroxyphenyl)-2-phenyl-3,5-pyrazolidinedione monohydrate, 4-Butyl-1-(p-hydroxyphenyl)-2-phenyl-3,5-pyrazolidinedione monohydrate, 1-(p-Hydroxyphenyl)-2-phenyl-4-butyl-3,5-pyrazolidinedione monohydrate, Oxyphenbutazone monohydrate, Tandearil monohydrate
InChI formel InChI=1S/C19H20N2O3.H2O/c1-2-3-9-17-18(23)20(14-7-5-4-6-8-14)21(19(17)24)15-10-12-16(22)13-11-15;/h4-8,10-13,17,22H,2-3,9H2,1H3;1H2
Løsningstype Neat
SMIL O.CCCCC1C(=O)N(N(C1=O)c2ccc(O)cc2)c3ccccc3
IUPAC navn 4-butyl-1-(4-hydroxyphenyl)-2-phenylpyrazolidine-3,5-dione;hydrate
Industry Type Clinical Testing
Molekylvægt (g/mol) 342.39
Mængde 10 mg
Formel vægt 342.158 g/mol