Få mere at vide
Oxypertine, LoGiCal
Artikelnummer. 30463629
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30463629 5 mg
1 options
Artikelnummer. 30463629 Leverandør LoGiCal Leverandørnr. LGCFOR3167.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxypertine
CAS 153-87-7
Analyt- eller komponentnavne Oxypertine
Forsendelsestilstand Room Temperature
Molekylær formel C23 H29 N3 O2
Certificeringer/compliance ISO 9001
Synonym 5,6-Dimethoxy-2-methyl-3-[2-(4-phenyl-1-piperazinyl)ethyl]-1H-indole, 1-[2-(5,6-Dimethoxy-2-methyl-3-indolyl)ethyl]-4-phenylpiperazine, 5,6-Dimethoxy-2-methyl-3-[2-(4-phenyl-1-piperazinyl)ethyl]indole, Equipertine, Forit, Integrin, Opertil, Oxypertin, Oxypertine, Win 18501-2
InChI formel InChI=1S/C23H29N3O2/c1-17-19(20-15-22(27-2)23(28-3)16-21(20)24-17)9-10-25-11-13-26(14-12-25)18-7-5-4-6-8-18/h4-8,15-16,24H,9-14H2,1-3H3
Løsningstype Neat
SMIL COc1cc2[nH]c(C)c(CCN3CCN(CC3)c4ccccc4)c2cc1OC
IUPAC navn 5,6-dimethoxy-2-methyl-3-[2-(4-phenylpiperazin-1-yl)ethyl]-1H-indole
Industry Type Clinical Testing
Molekylvægt (g/mol) 379.5
Mængde 5 mg
Formel vægt 379.226 g/mol