Få mere at vide
Oxymetazoline-d4, TRC
Artikelnummer. 30413074
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30413074 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413074 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8764721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxymetazoline dihydroimidazolyl-d4
CAS 1262053-60-0
Molekylær formel C16 2H4 H20 N2 O
Mængde 1 mg
Synonym Phenol','3-[(4,5-dihydro-4,5-d2-1H-imidazol-2-yl-4,5-d2)methyl]-6-(1,1-dimethylethyl)-2,4-dimethyl-, 3-[(4,5-Dihydro-4,5-d2-1H-imidazol-2-yl-4,5-d2)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol, Oxymetazoline-d4, Oxymetazoline-D4 (dihydroimidazolyl D4)
InChI formel InChI=1S/C16H24N2O/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14/h8,19H,6-7,9H2,1-5H3,(H,17,18)/i6D2,7D2
SMIL [2H]C1([2H])NC(=NC1([2H])[2H])Cc2c(C)cc(c(O)c2C)C(C)(C)C
IUPAC navn 6-tert-butyl-2,4-dimethyl-3-[(4,4,5,5-tetradeuterio-1H-imidazol-2-yl)methyl]phenol
Molekylvægt (g/mol) 264.4
Formel vægt 264.214