Få mere at vide
Oxybutynin N-Oxide 90%, TRC
Artikelnummer. 30426330
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426330 10 mg
30413775 5 mg
30420545 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426330 Leverandør Toronto Research Chemicals Leverandørnr. TRCO86853010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80976-68-7
Molekylær formel C22 H31 N O4
Molekylvægt (g/mol) 373.49
Synonym Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(diethyloxidoamino)-2-butyn-1-yl ester, Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(diethylamino)-2-butynyl ester, N-oxide, Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(diethyloxidoamino)-2-butynyl ester (9CI), Oxybutynin N-oxide, α-Cyclohexyl-α-hydroxybenzeneacetic acid 4-(diethylamino)-2-butynyl ester N-oxide, 4-Diethylamino-2-butynyl α-phenylcyclohexaneglycolate N-oxide
IUPAC navn 4-(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxy-N,N-diethylbut-2-yn-1-amine oxide
SMIL CC[N+]([O-])(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Oxybutynin N-Oxide
Mængde 10 mg
InChI formel InChI=1S/C22H31NO4/c1-3-23(26,4-2)17-11-12-18-27-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3
Formel vægt 373.2253