Få mere at vide
Oxybutynin EP Impurity D, Mikromol™
Artikelnummer. 30460292
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460292 4 x 25 mg
17954336 100 mg
17901386 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460292 Leverandør Mikromol Leverandørnr. MM0047.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxybutynin EP Impurity D, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenylcyclohexylglycolic Acid
CAS 4335-77-7
Analyt- eller komponentnavne Phenylcyclohexylglycolic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C14 H18 O3
Certificeringer/compliance ISO 17025
Synonym (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid (Phenylcyclohexylglycolic Acid), Oxybutynin Hydrochloride Imp. D (EP), Oxybutynin Imp. D (EP), Oxybutynin USP Related Compound A, Oxybutynin USP RC A, (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid, Phenylcyclohexylglycolic Acid, Oxybutynin Related Compound A, Oxybutynin Hydrochloride Impurity D, Oxybutynin Impurity D
InChI formel InChI=1S/C14H18O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12,17H,2,5-6,9-10H2,(H,15,16)
Løsningstype Neat
SMIL OC(=O)C(O)(C1CCCCC1)c2ccccc2
IUPAC navn 2-cyclohexyl-2-hydroxy-2-phenylacetic acid
Molekylvægt (g/mol) 234.29
Mængde 4 x 25 mg
Formel vægt 234.1256 g/mol