Få mere at vide
Oxybutynin-d10 Hydrochloride, TRC
Artikelnummer. 30414019
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414019 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414019 Leverandør Toronto Research Chemicals Leverandørnr. TRCO86852810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxybutynin-d10 Hydrochloride
CAS 120092-65-1
Molekylær formel C22H22D10ClNO3
Mængde 10 mg
Synonym α-Phenylcyclohexaneglycolic Acid 4-(Diethyl-d10-amino)-2-butynyl Ester','Hydrochloride
InChI formel InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H/i1D3,2D3,3D2,4D2;
SMIL OC(C(OCC#CCN(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H])=O)(C1=CC=CC=C1)C2CCCCC2.Cl
Molekylvægt (g/mol) 404.01
Formel vægt 403.2698