Få mere at vide
Oxybuprocaine Hydrochloride, Mikromol™
Beskrivelse
Oxybuprocaine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Oxybuprocaine Hydrochloride |
| CAS | 5987-82-6 |
| Analyt- eller komponentnavne | Oxybuprocaine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H28 N2 O3 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI,9CI), 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, Benoxil, Benoxinate hydrochloride, Boxinate, Cebesine, Conjucaine, Conjuncain, Dorsacaine, Dorsacaine hydrochloride, Lacrimin, Novesin, Novesine, Oxybuprocaine hydrochloride |
| InChI formel | InChI=1S/C17H28N2O3.ClH/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3;/h8-9,13H,4-7,10-12,18H2,1-3H3;1H |
| SMIL | Cl.CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC |
| IUPAC navn | 2-(diethylamino)ethyl 4-amino-3-butoxybenzoate;hydrochloride |
| Vis mere |