Få mere at vide
[Oxybis(methanetriyl)]tetrabenzene (Bis(diphenylmethyl) Ether), Mikromol™
Beskrivelse
[Oxybis(methanetriyl)]tetrabenzene (Bis(diphenylmethyl) Ether)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bis(diphenylmethyl) Ether |
| CAS | 574-42-5 |
| Analyt- eller komponentnavne | Bis(diphenylmethyl) Ether |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C26 H22 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Dimenhydrinate Imp. K (EP), Modafinil USP Related Compound D, Modafinil USP RC D, [Oxybis(methanetriyl)]tetrabenzene, Bis(diphenylmethyl) Ether, Modafinil Related Compound D, Dimenhydrinate Impurity K, Dimenhydrinate Impurity K, Ebastine Impurity H |
| InChI formel | InChI=1S/C26H22O/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22)27-26(23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20,25-26H |
| SMIL | O(C(c1ccccc1)c2ccccc2)C(c3ccccc3)c4ccccc4 |
| IUPAC navn | [benzhydryloxy(phenyl)methyl]benzene |
| Vis mere |