Få mere at vide
Oxprenolol Hydrochloride, Mikromol™
Beskrivelse
Oxprenolol Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Oxprenolol hydrochloride |
| CAS | 6452-73-9 |
| Analyt- eller komponentnavne | Oxprenolol hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H23 N O3 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yloxy)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyloxy)phenoxy]-, hydrochloride (9CI), 2-Propanol, 1-[o-(allyloxy)phenoxy]-3-(isopropylamino)-, hydrochloride (7CI,8CI), 1-(2-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(2'-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propanol hydrochloride, 1-(o-Allyloxyphenoxy)-3-isopropylamino-2-propranol-hydrochloride, 1-Isopropyl-amino-2-hydroxy-3-(O-allyloxyphenoxy)propane hydrochloride, 1-Isopropylamino-3-(o-allyloxyphenoxy)-2-propanol hydrochloride, Ba 39089, CIBA 39089Ba, Laracor, Oxiprenolol hydrochloride, Oxprenolol hydrochloride, Paritane, Slow-Pren, Slow-Trasicor, Trasacor, Trasicor, Trasicor 80, dl-Alprenolol hydrochloride, dl-Oxprenolol hydrochloride, (+/-)-1-[o-(Allyloxy)phenoxy]-3-isopropylamino-2-propanol hydrochloride, (+/-)-Ba 39089, (+/-)-Oxprenolol hydrochloride |
| InChI formel | InChI=1S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17H,1,9-11H2,2-3H3;1H |
| SMIL | Cl.CC(C)NCC(O)COc1ccccc1OCC=C |
| IUPAC navn | 1-(propan-2-ylamino)-3-(2-prop-2-enoxyphenoxy)propan-2-ol;hydrochloride |
| Vis mere |