Få mere at vide
Oxprenolol-d7 Hydrochloride, TRC
Artikelnummer. 30416216
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30416216 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416216 Leverandør Toronto Research Chemicals Leverandørnr. TRCO87050210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxprenolol-d7 Hydrochloride
CAS 1189649-47-5
Molekylær formel C15H17D7ClNO3
Mængde 10 mg
Synonym 1-[(1-Methylethyl-d7)amino]-3-[2-(2-propen-1-yloxy)phenoxy]-2-propanol Hydrochloride, 1-(Isopropylamino-d7)-2-hydroxy-3-[o-
InChI formel InChI=1S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17H,1,9-11H2,2-3H3;1H/i2D3,3D3,12D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(NCC(O)COc1ccccc1OCC=C)C([2H])([2H])[2H]
Molekylvægt (g/mol) 308.85
Formel vægt 308.1884