Få mere at vide
Oxozolpidem (N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide), Mikromol™
Artikelnummer. 17944356
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17944356 100 mg
17941496 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17944356 Leverandør Mikromol Leverandørnr. MM0333.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxozolpidem (N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxozolpidem (N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide)
CAS 400038-68-8
Analyt- eller komponentnavne Oxozolpidem (N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide)
Forsendelsestilstand Room Temperature
Molekylær formel C19 H19 N3 O2
Certificeringer/compliance ISO 17025
Synonym Oxozolpidem, N,N,6-Trimethyl-2-(4-methylphenyl)-alpha-oxo-imidazo[1,2-a]pyridine-3-acetamide, Zolpidem Related Compound B
InChI formel InChI=1S/C19H19N3O2/c1-12-5-8-14(9-6-12)16-17(18(23)19(24)21(3)4)22-11-13(2)7-10-15(22)20-16/h5-11H,1-4H3
SMIL CN(C)C(=O)C(=O)c1c(nc2ccc(C)cn12)c3ccc(C)cc3
IUPAC navn N,N-dimethyl-2-[6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]-2-oxoacetamide
Molekylvægt (g/mol) 321.37
Mængde 100 mg
Formel vægt 321.1477 g/mol