Learn More
Thermo Scientific™ Oxoid™ STA Selective Supplement
The Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
Brand:  Thermo Scientific™ SR0162E
Additional Details : Weight : 0.20000kg
Product Code. 11474209
  • 995.00 DKK / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
Specifications
Specifications
STA Selective Supplement
Selective Supplement
Each vial for 500mL medium
Freeze-dried
10/Pk.