Learn More
Thermo Scientific™ Oxoid™ STA Selective Supplement
The Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
Brand: Thermo Scientific™ SR0162E
Additional Details : Weight : 0.20000kg
Description
Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
- Add to STAA Agar Base, Part No. CM0881B
- Each vial supplements 500mL of medium
Specifications
STA Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |