missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs

Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.

Brand:  Thermo Scientific™ CT0522B

 View more versions of this product

Product Code. 11933822

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs.

Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.

disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mupirocin
5ÎĽg
MUP
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with BSAC, SFM guidelines
Oxoid Mupirocin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3783754 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog NumberCT0522B
Lot Number3636095 Certificate TypeCertificate of Analysis Date15/03/2023 Catalog NumberCT0522B
Lot Number3144972 Certificate TypeCertificate of Analysis Date07/10/2020 Catalog NumberCT0522B
Lot Number2468989 Certificate TypeCertificate of Analysis Date01/02/2019 Catalog NumberCT0522B
Lot Number1785737 Certificate TypeCertificate of Analysis Date11/07/2018 Catalog NumberCT0522B
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.