Learn More
Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 ÎĽg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Brand:  Thermo Scientific™ CT0758B
210.90 DKK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Product Code. 11902962
  • 228.00 DKK / Pack of 250
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks.
The disc contains 200ÎĽg of the active ingredient, fosfomycin, with the addition of 50ÎĽg of glucose-6-phospate, as required by the EUCAST methodology. Discs must be used for E.coli only and for other Enterobacterales and Staphylococcus spp., an MIC method must be used.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fosfomycin
Oxoid Fosfomycin Antimicrobial Susceptibility Disks
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
200ÎĽg
FOT
Antimicrobial Susceptibility
50 Disks/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6267567 Certificate TypeCertificate of Analysis Date21/05/2025 Catalog NumberCT0758B
Lot Number6267568 Certificate TypeCertificate of Analysis Date21/05/2025 Catalog NumberCT0758B
Lot Number6267566 Certificate TypeCertificate of Analysis Date21/05/2025 Catalog NumberCT0758B
Lot Number6259594 Certificate TypeCertificate of Analysis Date02/05/2025 Catalog NumberCT0758B
Lot Number6259593 Certificate TypeCertificate of Analysis Date02/05/2025 Catalog NumberCT0758B
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 ÎĽg >