missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cefixime Rhamnose Sorbitol MacConkey Agar Base (Dehydrated)

Selectively detect Escherichia coli O157 with this selective, differential medium.

Brand:  Thermo Scientific™ CM1005B

2334.96 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11973892

  • 2525.00 DKK / 500g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Selectively detect Escherichia coli O157 with Thermo Scientific™ Oxoid™ Cefixime Rhamnose Sorbitol MacConkey Agar Base (Dehydrated), a selective, differential medium based on Sorbitol MacConkey Agar with added rhamnose and cefixime. The addition of rhamnose to Sorbitol MacConkey Agar (SMAC) has been shown to aid the differentiation of Escherichia coli O157 from background flora.

Escherichia coli O157 is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhoea and severe abdominal pain, and hemolytic uraemic syndrome (HUS) 1,2,3,4,5, and as such, is a significant human pathogen.

Cefixime Rhamnose Sorbitol MacConkey Agar Base provides a selective base for differentiation of Escherichia coli O157.

  • Selective: the addition of Cefixime reduces the level of competing flora, particularly Proteus, that often account for large numbers of non-sorbitol fermenting colonies
  • Easy-to-read: Escherichia coli O157 do not usually ferment sorbitol or rhamnose, so appear as straw-colored colonies, while the sorbitol negative Escherichia coli that ferment rhamnose appear as pink colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cefixime Rhamnose Sorbitol MacConkey Agar Base
Agar
For 8.8L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Cefixime Rhamnose Sorbitol MacConkey Agar Base (Dehydrated) > Cefixime Rhamnose Sorbitol MacConkey Agar Base; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only