Learn More
Thermo Scientific™ Oxoid An-Ident Discs
Product Code. 13276049
Change view
Click to view available options
Quantity:
50 Disks
Unit Size:
Pack of 300
Product Code. Quantity Unit Size
13276049 50 Disks Pack of 300
1 options
Product Code. 13276049 Supplier Thermo Scientific™ Supplier No. DD0006A
  • ONLINE EXCLUSIVE PRICE
    710.00 DKK
    627.58 DKK / Pakke med 300
    Spare 82.42 DKK
    11% Slukket
 Restricted Item
Forventet forsendelse: 27-11-2026
Add to basket
Edge
Add to basket
Includes
  • Erythromycin, 60μg
  • Rifampin, 15μg
  • Colistin, 10μg
  • Penicillin, 2U
  • Kanamycin, 1000μg
  • Vancomycin, 5μg
Presumptive identification for Gram-negative anaerobic organisms
Order Info
Supplied 50 disks per cartridge; 6 cartridges/set.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Gram-negative anaerobic organisms
Type Oxoid An-Ident Discs
Includes Erythromycin 60μg, Rifampin 15μg, Colistin 10μg, Penicillin 2U, Kanamycin 1000μg, Vancomycin 5μg
Quantity 50 Disks