Få mere at vide
Oxo Sotalol-d6 Hydrochloride, TRC
Artikelnummer. 30426903
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30426903 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426903 Leverandør Toronto Research Chemicals Leverandørnr. TRCO87042710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxo Sotalol-d6 Hydrochloride
CAS 1346605-37-5
Molekylær formel C12 H12 D6 N2 O3 S • HCl
Mængde 10 mg
Synonym 4'-[(Isopropylamino)acetyl]methanesulfonanilide-d6 Hydrochloride, N-[4-[[(1-Methylethyl)amino]acetyl]phenyl]methanesulfonamide-d6 Hydrochloride
InChI formel InChI=1S/C12H18N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,13-14H,8H2,1-3H3;1H/i1D3,2D3;
SMIL [2H]C([2H])([2H])C(C([2H])([2H])[2H])NCC(=O)C1=CC=C(C=C1)NS(=O)(=O)C.Cl
IUPAC navn N-(4-((propan-2-yl-1,1,1,3,3,3-d6)glycyl)phenyl)methanesulfonamide hydrochloride
Molekylvægt (g/mol) 312.85
Formel vægt 312.12