Få mere at vide
Oxo-MDV 3100 (Oxo-enzalutamide), TRC
Artikelnummer. 30138015
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138015 10 mg
30132876 50 mg
30010420 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138015 Leverandør Toronto Research Chemicals Leverandørnr. TRCO85780010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1242137-18-3
Molekylær formel C21 H16 F4 N4 O3
Molekylvægt (g/mol) 448.37
Synonym 4-(3-(4-Cyano-3-(trifluoromethyl)phenyl)-5,5-dimethyl-2,4-dioxoimidazolidin-1-yl)-2-fluoro-N-methylbenzamide
IUPAC navn 4-[3-[4-cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-2,4-dioxo-imidazolidin-1-yl]-2-fluoro-N-methyl-benzamide
SMIL CNC(=O)c1ccc(cc1F)N2C(=O)N(C(=O)C2(C)C)c3ccc(C#N)c(c3)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Oxo Enzalutamide
Mængde 10 mg
InChI formel InChI=1S/C21H16F4N4O3/c1-20(2)18(31)28(12-5-4-11(10-26)15(8-12)21(23,24)25)19(32)29(20)13-6-7-14(16(22)9-13)17(30)27-3/h4-9H,1-3H3,(H,27,30)
Formel vægt 448.1159