Få mere at vide
Oxiconazole Nitrate, TRC
Artikelnummer. 30428112
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428112 100 mg
30428457 5 mg
30411133 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428112 Leverandør Toronto Research Chemicals Leverandørnr. TRCO846800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64211-46-7
Molekylær formel C18 H13 Cl4 N3 O . H N O3
Molekylvægt (g/mol) 492.14
Synonym Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (1Z)-, mononitrate (1:1), Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (1Z)-, mononitrate (9CI), Ethanone, 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)-, O-[(2,4-dichlorophenyl)methyl]oxime, (Z)-, mononitrate, Gyno-Myfungar, Myfungar, Oceral, Oxiconazole nitrate, Oxistat, Ro 13-8996, ST 813, Scorba LX, Sgd 301-76
IUPAC navn (Z)-1-(2,4-dichlorophenyl)-N-[(2,4-dichlorophenyl)methoxy]-2-imidazol-1-ylethanimine;nitric acid
SMIL O[N+](=O)[O-].Clc1ccc(CO\N=C(/Cn2ccnc2)\c3ccc(Cl)cc3Cl)c(Cl)c1
Tekniske data
Kemisk navn eller materiale Oxiconazole Nitrate
Mængde 100 mg
InChI formel InChI=1S/C18H13Cl4N3O.HNO3/c19-13-2-1-12(16(21)7-13)10-26-24-18(9-25-6-5-23-11-25)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11H,9-10H2;(H,2,3,4)/b24-18+;
Formel vægt 489.9769