Få mere at vide
Oxazole-4-carbohydrazide, TRC
Artikelnummer. 30383632
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30383632 100 mg
30373490 500 mg
30372809 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30383632 Leverandør Toronto Research Chemicals Leverandørnr. TRCE588903100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 885274-12-4
Molekylær formel C4H5N3O2
Molekylvægt (g/mol) 127.1
SMIL NNC(=O)c1cocn1
Tekniske data
Kemisk navn eller materiale Oxazole-4-carbohydrazide
Mængde 100 mg
InChI formel InChI=1S/C4H5N3O2/c5-7-4(8)3-1-9-2-6-3/h1-2H,5H2,(H,7,8)
Formel vægt 127.0382