Få mere at vide
Oxantel Pamoate, TRC
Artikelnummer. 17912185
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17912185 100 mg
17994235 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17912185 Leverandør Toronto Research Chemicals Leverandørnr. TRCO845250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68813-55-8
Molekylær formel C23 H16 O6 . C13 H16 N2 O
Molekylvægt (g/mol) 604.65
Synonym 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with (E)-3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), Phenol, 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt) (9CI), Phenol, 3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, (E)-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt), CP 14445-16, Oxantel ebonate, Oxantel pamoate, Telopar
IUPAC navn 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;3-[(E)-2-(1-methyl-5,6-dihydro-4H-pyrimidin-2-yl)ethenyl]phenol
SMIL CN1CCCN=C1\C=C\c2cccc(O)c2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
Tekniske data
Kemisk navn eller materiale Oxantel Pamoate
Mængde 100 mg
InChI formel InChI=1S/C23H16O6.C13H16N2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-15-9-3-8-14-13(15)7-6-11-4-2-5-12(16)10-11/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-7,10,16H,3,8-9H2,1H3/b;7-6+
Formel vægt 604.221