Få mere at vide
Oxaliplatin-d10, TRC
Artikelnummer. 30410614
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30410614 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410614 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8450772.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oxaliplatin-d10
CAS 1132819-16-9
Molekylær formel C8H4D10N2O4Pt
Mængde 2.5 mg
Synonym [SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N')[ethanedioato(2-)-O,O'] platinum-d10, (SP-4-2)-[(1R,2R)-1,2-Cyclohexanediamine-kN,kN'][ethanedioato(2-)-kO1,kO2]platinum, (1R-trans)-1,2-Cyclohexanediamine Platinum Complex-d10, Dacplat-d10, Eloxatin-d10, Elplat-d10, Lipoxal-d10, Oxalatoplatinum-d10
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])[C@]2([2H])[C@@]([2H])(C1([2H])[2H])[NH2][Pt]3(OC(=O)C(=O)O3)[NH2]2
IUPAC navn (1'R,6'R)-1',2',2',3',3',4',4',5',5',6'-decadeuteriospiro[1,3-dioxa-2λ^{4}-platinacyclopentane-2,8'-7λ^{4},9λ^{4}-diaza-8λ^{4}-platinabicyclo[4.3.0]nonane]-4,5-dione
Molekylvægt (g/mol) 407.35
Formel vægt 407.1229