Få mere at vide
Oxacillin Sodium Monohydrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Oxacillin sodium salt hydrate |
| CAS | 7240-38-2 |
| Analyt- eller komponentnavne | Oxacillin sodium salt hydrate |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C19 H18 N3 O5 S . Na . H2O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Oxacillin Sodium Monohydrate, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-[[(5-methyl-3-phenyl-4-isoxazolyl)carbonyl]amino]-7-oxo-, sodium salt, hydrate (1:1:1), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-(5-methyl-3-phenyl-4-isoxazolecarboxamido)-7-oxo-, monosodium salt, monohydrate (8CI), Oxacillin sodium hydrate, Sodium oxacillin monohydrate, Stapenor sodium, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-[[(5-methyl-3-phenyl-4-isoxazolyl)carbonyl]amino]-7-oxo-, monosodium salt, monohydrate, [2S-(2alpha,5alpha,6beta)]- (9CI) |
| InChI formel | InChI=1S/C19H19N3O5S.Na.H2O/c1-9-11(12(21-27-9)10-7-5-4-6-8-10)15(23)20-13-16(24)22-14(18(25)26)19(2,3)28-17(13)22;;/h4-8,13-14,17H,1-3H3,(H,20,23)(H,25,26);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1 |
| Løsningstype | Neat |
| SMIL | O.[Na+].Cc1onc(c2ccccc2)c1C(=O)N[C@H]3[C@H]4SC(C)(C)[C@@H](N4C3=O)C(=O)[O-] |
| Vis mere |